The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019821.2 | NP_062795.2 | 1 - 2 | 173 | 85 |
GenBank mRNA | AF209703.1 | - | 1 - 2 | 99 | 85 |
AK157313.1 | - | 1 - 2 | 160 | 85 | |
BC016584.1 | - | 1 - 2 | 173 | 85 | |
BC053729.1 | - | 1 - 2 | 116 | 85 |