Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nell1
Entrez Gene ID
338352
Gene Name
NEL-like 1
Gene Aliases
B230343H07Rik, l7R6
Chromosome Location
Chr.7: 49975350 - 50863289 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037906.2 NP_001032995.1 4 - 5 542 69
GenBank mRNA AY622226.1 - 4 - 5 542 69


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK035921.1 - 1031 Chr. - - [-] on Build 69

View Full Product Details