The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008422.1 | NP_001008422.1 | 2 - 3 | 289 | 102 |
GenBank mRNA | AK141920.1 | - | 3 - 4 | 303 | 102 |
BC042576.1 | - | 2 - 3 | 147 | 102 | |
BC043138.1 | - | 2 - 3 | 319 | 102 | |
BC054818.1 | - | 1 - 2 | 499 | 102 | |
BC079873.1 | - | 2 - 3 | 307 | 102 | |
BC085153.1 | - | 2 - 3 | 289 | 102 |