The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077591.1 | NP_001071059.1 | 7 - 8 | 1185 | 65 |
NM_011651.2 | NP_035781.2 | 7 - 8 | 1212 | 65 | |
GenBank mRNA | BC118618.1 | - | 7 - 8 | 1164 | 65 |
BC121827.1 | - | 7 - 8 | 1164 | 65 |