The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033371.2 | NP_203535.1 | 3 - 4 | 1140 | 75 |
GenBank mRNA | AK154865.1 | - | 3 - 4 | 1125 | 75 |
AK171452.1 | - | 3 - 4 | 1125 | 75 | |
AK220248.1 | - | 3 - 4 | 1140 | 75 | |
AY010723.1 | - | 2 - 3 | 335 | 75 | |
BC033318.1 | - | 2 - 3 | 700 | 75 | |
BC044775.1 | - | 2 - 3 | 822 | 75 | |
BC049958.1 | - | 2 - 3 | 822 | 75 | |
BC057450.1 | - | 2 - 3 | 757 | 75 | |
BC096620.1 | - | 3 - 4 | 1140 | 75 |