The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198019.2 | NP_932136.2 | 1 - 2 | 374 | 87 |
GenBank mRNA | AK050870.1 | - | 1 - 2 | 81 | 87 |
BC070400.1 | - | 1 - 2 | 374 | 87 | |
BC071270.1 | - | 1 - 2 | 358 | 87 |