The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178891.5 | NP_849222.3 | 1 - 1 | 2338 | 104 |
GenBank mRNA | AK034732.1 | - | 1 - 1 | 2308 | 104 |
AK172003.1 | - | 1 - 1 | 2330 | 104 | |
BG229470.1 | - | 1 - 1 | 173 | 104 |