Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Usp9y
Entrez Gene ID
107868
Gene Name
ubiquitin specific peptidase 9, Y chromosome
Gene Aliases
Dffry, Fafl2
Chromosome Location
Chr.Y: 1298961 - 1459782 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_148943.2 NP_683745.2 40 - 41 6998 114
GenBank mRNA AJ307017.1 - 40 - 41 6998 114

View Full Product Details