Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gnpda2
Entrez Gene ID
67980
Gene Name
glucosamine-6-phosphate deaminase 2
Gene Aliases
4921523I18Rik, 4933412A11Rik, BB189630, GNPDA, Gnp2, Sb52, mKIAA4008
Chromosome Location
Chr.5: 69574998 - 69592332 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038015.1 NP_001033104.1 3 - 4 343 142
GenBank mRNA AK014950.1 - 3 - 4 353 142
AK015527.1 - 3 - 4 353 142
AK016785.1 - - 398 142
AK017588.1 - 3 - 4 352 142
AK051596.1 - 3 - 4 352 142
AK141408.1 - 3 - 4 355 142
AK166601.1 - 3 - 4 345 142
AK168516.1 - 3 - 4 362 142
AK171299.1 - 3 - 4 338 142
AK220275.1 - 3 - 4 304 142
BC004084.1 - 3 - 4 343 142

View Full Product Details