Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nkain3
Entrez Gene ID
269513
Gene Name
Na+/K+ transporting ATPase interacting 3
Gene Aliases
C530048M05, E130310K16Rik
Chromosome Location
Chr.4: 20118336 - 20780310 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290410.1 NP_001277339.1 1 - 2 408 63
NM_172987.2 NP_766575.2 1 - 2 408 63
GenBank mRNA AK053823.1 - 1 - 2 293 63
AK141456.1 - 1 - 2 308 63
BC075683.1 - 1 - 2 408 63
EF058052.1 - 1 - 2 55 63
EF058053.1 - 1 - 2 55 63
EF058054.1 - 1 - 2 55 63

View Full Product Details