The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024248.1 | NP_077210.1 | 14 - 15 | 1679 | 67 |
GenBank mRNA | AK009937.1 | - | 14 - 15 | 1641 | 67 |
AK038968.1 | - | 14 - 15 | 1689 | 67 | |
BF458069.1 | - | 1 - 2 | 67 |