Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cars2
Entrez Gene ID
71941
Gene Name
cysteinyl-tRNA synthetase 2 (mitochondrial)(putative)
Gene Aliases
2310051N18Rik, 2410044A07Rik, D530030H10Rik
Chromosome Location
Chr.8: 11514012 - 11550778 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024248.1 NP_077210.1 7 - 8 839 77
GenBank mRNA AK009937.1 - 7 - 8 801 77
AK038968.1 - 7 - 8 849 77
AK041962.1 - 7 - 8 810 77
AK054486.1 - 7 - 8 833 77

View Full Product Details