The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001294138.1 | NP_001281067.1 | 7 - 8 | 953 | 124 |
NM_001294140.1 | NP_001281069.1 | 6 - 7 | 869 | 124 | |
NM_008194.3 | NP_032220.1 | 6 - 7 | 869 | 124 | |
NM_212444.2 | NP_997609.1 | 6 - 7 | 869 | 124 | |
GenBank mRNA | AK044308.1 | - | 6 - 7 | 707 | 124 |
AK088373.1 | - | 6 - 7 | 688 | 124 | |
AK169424.1 | - | 7 - 8 | 759 | 124 | |
AK169525.1 | - | 6 - 7 | 870 | 124 | |
AK201906.1 | - | 1 - 2 | 74 | 124 | |
BC003767.1 | - | 6 - 7 | 672 | 124 | |
U48403.1 | - | 6 - 7 | 646 | 124 |