The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113389.1 | NP_001106860.1 | 6 - 7 | 815 | 143 |
NM_007890.2 | NP_031916.1 | 6 - 7 | 1074 | 143 | |
GenBank mRNA | BC034550.1 | - | 1 - 2 | 160 | 143 |
BC129888.1 | - | 5 - 6 | 677 | 143 | |
BC129889.1 | - | 5 - 6 | 677 | 143 | |
BC138716.1 | - | 5 - 6 | 704 | 143 | |
BC145405.1 | - | 5 - 6 | 677 | 143 | |
U58497.1 | - | 5 - 6 | 704 | 143 |