Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Dnah12
- Entrez Gene ID
-
110083
- Gene Name
- dynein, axonemal, heavy chain 12
- Gene Aliases
-
4921531P07Rik, DHC3, DLP12, Dnah12l, Dnahc12, Dnahc3, Dnahc7c, Dnahc7l, Gm284, Gm74, Gm907, HL-19, HL19, Hdhc3
- Chromosome Location
-
Chr.14: 26691423 - 26892284 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK076605.1 |
- |
10 - 11 |
1296 |
70 |
View Full Product Details