Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Appl1
Entrez Gene ID
72993
Gene Name
adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 1
Gene Aliases
2900057D21Rik, 7330406P05Rik, AI585782, AW209077, BB022931, C88264, DIP13
Chromosome Location
Chr.14: 26918986 - 26971205 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145221.2 NP_660256.1 16 - 17 1568 90
GenBank mRNA AK030445.1 - 16 - 17 1447 90
AK033566.1 - 16 - 17 1539 90
AK045438.1 - 16 - 17 1504 90
AK049307.1 - 16 - 17 1555 90
AK146356.1 - 16 - 17 1539 90
AY113705.1 - 16 - 17 1533 90
BC019708.1 - 8 - 9 846 90
BC063751.1 - 16 - 17 1580 90

View Full Product Details