Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Il10rb
Entrez Gene ID
16155
Gene Name
interleukin 10 receptor, beta
Gene Aliases
6620401D04Rik, AI528744, CRF2-4, Crfb4, D16H21S58, D21S58h, IL-10R2, Il10r2
Chromosome Location
Chr.16: 91406235 - 91425834 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008349.5 NP_032375.2 2 - 3 254 69
GenBank mRNA AF440787.1 - 2 - 3 181 69
AK040938.1 - 2 - 3 170 69
AK134042.1 - 2 - 3 247 69
AK160991.1 - 2 - 3 1209 69
BC145791.1 - 2 - 3 254 69
U53696.1 - 2 - 3 167 69

View Full Product Details