Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Uty
Entrez Gene ID
22290
Gene Name
ubiquitously transcribed tetratricopeptide repeat gene, Y chromosome
Gene Aliases
Hydb, mKIAA4057
Chromosome Location
Chr.Y: 1085187 - 1245754 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009484.2 NP_033510.2 9 - 10 921 89
GenBank mRNA AF057367.1 - 9 - 10 901 89
AK043154.1 - 7 - 8 741 89
AK142512.1 - 9 - 10 837 89
AK155922.1 - 10 - 11 801 89
AK162410.1 - 8 - 9 629 89
AK220463.1 - 10 - 11 1009 89
BC053061.1 - 9 - 10 904 89

View Full Product Details