Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ept1
Entrez Gene ID
28042
Gene Name
ethanolaminephosphotransferase 1 (CDP-ethanolamine-specific)
Gene Aliases
4933402G07Rik, AI448296, AI452230, C79563, D5Wsu178e, SELI, mKIAA1724
Chromosome Location
Chr.5: 30232618 - 30272430 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027652.2 NP_081928.2 6 - 7 822 136
GenBank mRNA AK040745.1 - 6 - 7 732 136
AK048011.1 - 6 - 7 771 136
AK079325.1 - 6 - 7 798 136
AK122544.1 - 6 - 7 738 136
BC106097.1 - 6 - 7 791 136
BC115783.1 - 6 - 7 761 136
BC117551.1 - 6 - 7 756 136

View Full Product Details