Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ept1
Entrez Gene ID
28042
Gene Name
ethanolaminephosphotransferase 1 (CDP-ethanolamine-specific)
Gene Aliases
4933402G07Rik, AI448296, AI452230, C79563, D5Wsu178e, SELI, mKIAA1724
Chromosome Location
Chr.5: 30232618 - 30272430 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027652.2 NP_081928.2 7 - 8 875 91
GenBank mRNA AK014530.1 - 6 - 7 687 91
AK040745.1 - 7 - 8 785 91
AK048011.1 - 7 - 8 824 91
AK079325.1 - 7 - 8 851 91
AK122544.1 - 7 - 8 791 91
BC106097.1 - 7 - 8 844 91
BC115783.1 - 7 - 8 814 91
BC117551.1 - 7 - 8 809 91

View Full Product Details