Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Necab1
Entrez Gene ID
69352
Gene Name
N-terminal EF-hand calcium binding protein 1
Gene Aliases
1700003H21Rik, Efcbp1, STIP-1
Chromosome Location
Chr.4: 14952245 - 15149804 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178617.4 NP_848732.3 2 - 3 340 111
GenBank mRNA AK039240.1 - 2 - 3 156 111
AK050307.1 - 3 - 4 237 111
AK081951.1 - 2 - 3 232 111
AY278200.2 - 2 - 3 323 111
BC067055.1 - 2 - 3 340 111

View Full Product Details