Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Schip1
Entrez Gene ID
30953
Gene Name
schwannomin interacting protein 1
Gene Aliases
Nf2ip, Schip-1
Chromosome Location
Chr.3: 68064802 - 68626482 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113420.1 NP_001106891.1 6 - 7 863 69
NM_001113421.1 NP_001106892.1 7 - 8 1547 69
NM_001282044.1 NP_001268973.1 7 - 8 1530 69
NM_001282045.1 NP_001268974.1 7 - 8 1388 69
NM_013928.5 NP_038956.2 6 - 7 953 69
GenBank mRNA AF145716.1 - 6 - 7 1059 69
AK021361.1 - 6 - 7 814 69
AK083464.1 - 7 - 8 1373 69
AK146930.1 - 7 - 8 1504 69
AK168005.1 - 7 - 8 1457 69
BC060529.1 - 6 - 7 887 69
EU163407.1 - 7 - 8 1547 69
EU163408.1 - 6 - 7 863 69
KM233715.1 - 6 - 7 809 69
KM233716.1 - 6 - 7 814 69

Gene Symbol
Gm21949
Entrez Gene ID
100505386
Gene Name
predicted gene, 21949
Gene Aliases
Iqcj-schip1, Schip-1-iqcj, Schip1
Chromosome Location
Chr.3: 67892220 - 68626482 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113419.2 NP_001106890.1 10 - 11 1725 69
GenBank mRNA KJ941154.1 - 9 - 10 1029 69

View Full Product Details