Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ap3m2
Entrez Gene ID
64933
Gene Name
adaptor-related protein complex 3, mu 2 subunit
Gene Aliases
5830445E16Rik, AP-3B
Chromosome Location
Chr.8: 22787354 - 22805654 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122820.1 NP_001116292.1 7 - 8 1219 101
NM_029505.3 NP_083781.2 7 - 8 1157 101
GenBank mRNA AK077365.1 - 7 - 8 1134 101
AK081220.1 - 7 - 8 1126 101
AK082988.1 - 7 - 8 1115 101
AK134634.1 - 7 - 8 1169 101
AK162771.1 - 2 - 3 262 101
BC027301.1 - 7 - 8 1103 101
BC030484.1 - 7 - 8 1141 101

View Full Product Details