Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fcer2a
Entrez Gene ID
14128
Gene Name
Fc receptor, IgE, low affinity II, alpha polypeptide
Gene Aliases
CD23, Fce2, Fcer2, Ly-42
Chromosome Location
Chr.8: 3680726 - 3694174 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001253737.1 NP_001240666.1 5 - 6 349 67
NM_001253739.1 NP_001240668.1 5 - 6 360 67
NM_001253743.1 NP_001240672.1 6 - 7 409 67
NM_001253745.1 NP_001240674.1 4 - 5 286 67
NM_001253747.1 NP_001240676.1 5 - 6 370 67
NM_013517.4 NP_038545.1 6 - 7 490 67
GenBank mRNA AK156654.1 - 6 - 7 493 67
AK172591.1 - 6 - 7 493 67
AY069980.1 - 4 - 5 252 67
BC119164.1 - 5 - 6 357 67
BC119190.1 - 5 - 6 357 67
M34163.1 - 6 - 7 492 67
M99371.1 - 6 - 7 482 67
S67072.1 - 5 - 6 341 67
S67073.1 - 4 - 5 221 67
S67074.1 - 5 - 6 260 67
X64223.1 - 5 - 6 349 67
X64224.1 - 5 - 6 360 67

View Full Product Details