The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159573.1 | NP_001153045.1 | 1 - 2 | 279 | 145 |
NM_001159574.1 | NP_001153046.1 | 1 - 2 | 279 | 145 | |
NM_024183.5 | NP_077145.2 | 1 - 2 | 279 | 145 | |
GenBank mRNA | AK005061.1 | - | 1 - 2 | 248 | 145 |
AK008561.1 | - | 1 - 2 | 251 | 145 | |
AK049672.1 | - | 1 - 2 | 265 | 145 | |
AK164105.1 | - | 1 - 2 | 245 | 145 | |
AK168464.1 | - | 1 - 2 | 280 | 145 | |
BC003263.1 | - | 1 - 2 | 237 | 145 |