The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009057.3 | NP_033083.2 | 5 - 6 | 674 | 55 |
GenBank mRNA | AK014305.1 | - | 5 - 6 | 627 | 55 |
AK045924.1 | - | 3 - 4 | 987 | 55 | |
AK079716.1 | - | 4 - 5 | 739 | 55 | |
AK158862.1 | - | 4 - 5 | 699 | 55 | |
AK170162.1 | - | 5 - 6 | 625 | 55 | |
BC014292.1 | - | 5 - 6 | 658 | 55 |