The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146131.2 | NP_666243.1 | 10 - 11 | 2200 | 75 |
GenBank mRNA | AK028754.1 | - | 10 - 11 | 2162 | 75 |
AK028858.1 | - | 10 - 11 | 2198 | 75 | |
AK047898.1 | - | 1 - 2 | 45 | 75 | |
AK081345.1 | - | 1 - 2 | 45 | 75 | |
AK159422.1 | - | 10 - 11 | 2187 | 75 | |
AK159448.1 | - | 10 - 11 | 2187 | 75 | |
AK159633.1 | - | 10 - 11 | 2177 | 75 | |
AK160103.1 | - | 10 - 11 | 2181 | 75 | |
AK170318.1 | - | 10 - 11 | 2156 | 75 | |
BC026838.1 | - | 9 - 10 | 2135 | 75 | |
BC027771.1 | - | 10 - 11 | 2170 | 75 | |
BC034200.1 | - | 10 - 11 | 2141 | 75 |