The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013587.2 | NP_038615.2 | 7 - 8 | 1023 | 79 |
GenBank mRNA | AJ639614.1 | - | 7 - 8 | 1030 | 79 |
AK089250.1 | - | 7 - 8 | 1026 | 79 | |
AK145710.1 | - | 7 - 8 | 1066 | 79 | |
AK150451.1 | - | 7 - 8 | 1036 | 79 | |
AK152062.1 | - | 1 - 2 | 85 | 79 | |
AK152485.1 | - | 7 - 8 | 1036 | 79 | |
AK153323.1 | - | 7 - 8 | 1036 | 79 | |
AK166616.1 | - | 7 - 8 | 1026 | 79 | |
AK166746.1 | - | 7 - 8 | 1037 | 79 | |
AK166819.1 | - | 7 - 8 | 1041 | 79 | |
AK168421.1 | - | 7 - 8 | 1026 | 79 | |
AK169132.1 | - | 7 - 8 | 1031 | 79 | |
AK171471.1 | - | 7 - 8 | 1026 | 79 | |
BC032170.1 | - | 7 - 8 | 1026 | 79 | |
BC046641.1 | - | 7 - 8 | 1015 | 79 | |
BC057979.1 | - | 7 - 8 | 994 | 79 | |
BC059887.1 | - | 7 - 8 | 840 | 79 | |
BC094324.1 | - | 7 - 8 | 1023 | 79 | |
D00622.1 | - | 7 - 8 | 1016 | 79 | |
S67967.1 | - | 7 - 8 | 1049 | 79 |