Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Acox3
Entrez Gene ID
80911
Gene Name
acyl-Coenzyme A oxidase 3, pristanoyl
Gene Aliases
BI685180, EST-s59, PCOX
Chromosome Location
Chr.5: 35582967 - 35614210 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030721.2 NP_109646.2 13 - 14 1613 81
GenBank mRNA AJ278430.1 - 13 - 14 1646 81
AK028818.1 - 13 - 14 2034 81
AK053960.1 - 13 - 14 1610 81
AK154112.1 - 12 - 13 1704 81
AK171598.1 - - 1596 81
BC044725.2 - 13 - 14 1633 81
BC055019.1 - 13 - 14 1613 81

View Full Product Details