Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Itih1
Entrez Gene ID
16424
Gene Name
inter-alpha trypsin inhibitor, heavy chain 1
Gene Aliases
ITI-HC1, Intin1, Itih-1
Chromosome Location
Chr.14: 30929180 - 30943321 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008406.3 NP_032432.2 11 - 12 1423 98
GenBank mRNA AK149500.1 - 11 - 12 1424 98
BC013465.1 - 11 - 12 1407 98
BC028814.1 - 11 - 12 1423 98
X70391.1 - 11 - 12 1403 98

View Full Product Details