Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Nek5
- Entrez Gene ID
-
330721
- Gene Name
- NIMA (never in mitosis gene a)-related expressed kinase 5
- Gene Aliases
-
6530411J22, BB049667
- Chromosome Location
-
Chr.8: 22073615 - 22125415 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK165108.1 |
- |
13 - 14 |
1189 |
74 |
View Full Product Details