Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Kcne2
- Entrez Gene ID
-
246133
- Gene Name
- potassium voltage-gated channel, Isk-related subfamily, gene 2
- Gene Aliases
-
2200002I16Rik, AW048273, MiRP1
- Chromosome Location
-
Chr.16: 92292389 - 92298137 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC022699.1 |
- |
1 - 2 |
76 |
90 |
View Full Product Details