The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009827.2 | NP_033957.1 | 4 - 5 | 1094 | 146 |
GenBank mRNA | AK004730.2 | - | 4 - 5 | 952 | 146 |
AK143895.1 | - | 4 - 5 | 942 | 146 | |
AK146738.1 | - | 4 - 5 | 1094 | 146 | |
AK147052.1 | - | 4 - 5 | 1094 | 146 | |
AK164301.1 | - | - | 686 | 146 | |
BC020534.1 | - | 4 - 5 | 914 | 146 |