Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rnf6
Entrez Gene ID
74132
Gene Name
ring finger protein (C3H2C3 type) 6
Gene Aliases
1200013I08Rik, 5730419H05Rik, AA537053
Chromosome Location
Chr.5: 146209193 - 146222191 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256085.1 NP_001243014.1 2 - 3 112 109
NM_001256087.1 NP_001243016.1 2 - 3 244 109
NM_028774.3 NP_083050.1 2 - 3 318 109
GenBank mRNA AK004745.1 - 2 - 3 298 109
AK029912.1 - 2 - 3 99 109
AK150269.1 - 2 - 3 99 109
AK152106.1 - 2 - 3 99 109
AY039004.1 - 2 - 3 218 109
BC138545.1 - 3 - 4 224 109
CX237430.1 - 2 - 3 244 109

View Full Product Details