The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199242.1 | NP_001186171.1 | 3 - 4 | 494 | 118 |
NM_001199243.1 | NP_001186172.1 | 2 - 3 | 392 | 118 | |
NM_001199244.1 | NP_001186173.1 | 2 - 3 | 535 | 118 | |
NM_001199245.1 | NP_001186174.1 | 2 - 3 | 388 | 118 | |
NM_030265.3 | NP_084541.3 | 2 - 3 | 320 | 118 | |
GenBank mRNA | AF305071.1 | - | 2 - 3 | 291 | 118 |
AF453243.1 | - | 2 - 3 | 228 | 118 | |
AK039048.1 | - | 2 - 3 | 321 | 118 | |
AK049662.1 | - | 2 - 3 | 272 | 118 | |
AK133416.1 | - | 2 - 3 | 240 | 118 | |
AK148685.1 | - | 2 - 3 | 537 | 118 | |
AK165773.1 | - | 2 - 3 | 262 | 118 | |
AY647240.1 | - | 2 - 3 | 271 | 118 | |
BC051130.1 | - | 3 - 4 | 494 | 118 | |
DQ148501.1 | - | 3 - 4 | 494 | 118 | |
DQ148502.1 | - | 1 - 2 | 252 | 118 | |
DQ148503.1 | - | 2 - 3 | 272 | 118 | |
DQ148504.1 | - | 2 - 3 | 388 | 118 |