Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fcer2a
Entrez Gene ID
14128
Gene Name
Fc receptor, IgE, low affinity II, alpha polypeptide
Gene Aliases
CD23, Fce2, Fcer2, Ly-42
Chromosome Location
Chr.8: 3680726 - 3694174 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001253747.1 NP_001240676.1 2 - 3 192 76
GenBank mRNA S67073.1 - 1 - 2 43 76

View Full Product Details