Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Letm1
Entrez Gene ID
56384
Gene Name
leucine zipper-EF-hand containing transmembrane protein 1
Gene Aliases
-
Chromosome Location
Chr.5: 33739675 - 33782704 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019694.1 NP_062668.1 5 - 6 1029 72
GenBank mRNA AB041582.1 - 5 - 6 1042 72
AF061026.1 - 5 - 6 1029 72
AK133849.1 - 5 - 6 1048 72
BC023862.1 - 5 - 6 1026 72
BC046326.1 - 5 - 6 1036 72
BC055865.1 - 5 - 6 1047 72
BC061115.1 - 5 - 6 1042 72
BC061690.1 - 5 - 6 1029 72

View Full Product Details