The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027276.2 | NP_081552.2 | 4 - 5 | 356 | 57 |
GenBank mRNA | AK147938.1 | - | 4 - 5 | 371 | 57 |
AK158014.1 | - | 5 - 6 | 629 | 57 | |
AK168978.1 | - | 4 - 5 | 343 | 57 | |
BC026606.1 | - | 4 - 5 | 356 | 57 |