Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Cars2
- Entrez Gene ID
-
71941
- Gene Name
- cysteinyl-tRNA synthetase 2 (mitochondrial)(putative)
- Gene Aliases
-
2310051N18Rik, 2410044A07Rik, D530030H10Rik
- Chromosome Location
-
Chr.8: 11514012 - 11550778 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK054486.1 |
- |
9 - 10 |
1034 |
110 |
View Full Product Details