Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ept1
- Entrez Gene ID
-
28042
- Gene Name
- ethanolaminephosphotransferase 1 (CDP-ethanolamine-specific)
- Gene Aliases
-
4933402G07Rik, AI448296, AI452230, C79563, D5Wsu178e, SELI, mKIAA1724
- Chromosome Location
-
Chr.5: 30232618 - 30272430 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK014530.1 |
- |
3 - 4 |
320 |
62 |
View Full Product Details