Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mlf1
Entrez Gene ID
17349
Gene Name
myeloid leukemia factor 1
Gene Aliases
HLS7
Chromosome Location
Chr.3: 67374097 - 67400000 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039543.2 NP_001034632.1 2 - 3 316 77
GenBank mRNA BC027526.1 - 2 - 3 265 77

View Full Product Details