Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Khk
Entrez Gene ID
16548
Gene Name
ketohexokinase
Gene Aliases
-
Chromosome Location
Chr.5: 30921436 - 30931248 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008439.3 NP_032465.2 4 - 5 437 59
GenBank mRNA BC013464.1 - 4 - 5 437 59
Y09335.1 - 4 - 5 416 59

View Full Product Details