Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Katnal1
Entrez Gene ID
231912
Gene Name
katanin p60 subunit A-like 1
Gene Aliases
-
Chromosome Location
Chr.5: 148871584 - 148929134 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153572.2 NP_705800.1 8 - 9 1143 81
GenBank mRNA AK049620.1 - 8 - 9 1497 81
BC030434.1 - 8 - 9 1130 81

View Full Product Details