The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021720.1 | NP_068366.1 | 3 - 4 | 600 | 104 |
GenBank mRNA | AK042415.1 | - | 3 - 4 | 625 | 104 |
AK080299.1 | - | 3 - 4 | 638 | 104 | |
AK132269.1 | - | 3 - 4 | 624 | 104 | |
BC043316.1 | - | 3 - 4 | 600 | 104 |