Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttc24
Entrez Gene ID
214191
Gene Name
tetratricopeptide repeat domain 24
Gene Aliases
A430025D11Rik
Chromosome Location
Chr.3: 88069410 - 88081987 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172526.3 NP_766114.1 6 - 7 1278 58
GenBank mRNA AK039884.1 - 6 - 7 1279 58
BC147554.1 - 5 - 6 982 58
BC147555.1 - 5 - 6 982 58

View Full Product Details