The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172526.3 | NP_766114.1 | 9 - 10 | 1547 | 62 |
GenBank mRNA | AK039884.1 | - | 9 - 10 | 1548 | 62 |
AK076743.1 | - | 4 - 5 | 363 | 62 | |
BC055114.1 | - | 4 - 5 | 362 | 62 | |
BC147554.1 | - | 8 - 9 | 1251 | 62 | |
BC147555.1 | - | 8 - 9 | 1251 | 62 |