Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc7a1
Entrez Gene ID
11987
Gene Name
solute carrier family 7 (cationic amino acid transporter, y+ system), member 1
Gene Aliases
4831426K01Rik, AI447493, Atrc-1, Atrc1, Cat1, Rec-1, Rev-1, mCAT-1
Chromosome Location
Chr.5: 148327410 - 148399904 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301424.1 NP_001288353.1 12 - 13 1929 65
NM_007513.4 NP_031539.3 12 - 13 2045 65
GenBank mRNA AK139342.1 - 12 - 13 2021 65
AK147991.1 - 12 - 13 1954 65
BC068234.1 - 12 - 13 1936 65
BC145779.1 - 11 - 12 1850 65
BC145781.1 - 11 - 12 1850 65
JN226407.1 - 10 - 11 1765 65
JN226408.1 - 10 - 11 1765 65
JN226410.1 - 10 - 11 1765 65
M26687.1 - 12 - 13 1963 65

View Full Product Details