Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fcrls
Entrez Gene ID
80891
Gene Name
Fc receptor-like S, scavenger receptor
Gene Aliases
2810439C17Rik, 9330158F12, FcRH2sc, Fcrh2, IFGP2, IgSR, MMAN-g, Msr2, moFcRH2sc
Chromosome Location
Chr.3: 87250758 - 87263765 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030707.3 NP_109632.2 7 - 8 1553 85
GenBank mRNA AB028174.1 - 7 - 8 1584 85
AF302046.1 - 7 - 8 1622 85
AF329486.1 - 7 - 8 1554 85
AK034125.1 - 7 - 8 1554 85
AK076123.1 - 7 - 8 1587 85
AY506556.1 - 7 - 8 1593 85
BC016551.1 - 7 - 8 1596 85

View Full Product Details