Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spata18
Entrez Gene ID
73472
Gene Name
spermatogenesis associated 18
Gene Aliases
1700067I02Rik
Chromosome Location
Chr.5: 73651380 - 73679484 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178387.3 NP_848474.2 2 - 3 393 65
GenBank mRNA AK006916.1 - 1 - 2 60 65
BC050799.1 - 2 - 3 391 65

View Full Product Details