The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205355.1 | NP_001192284.1 | 1 - 2 | 147 | 101 |
GenBank mRNA | AK173244.1 | - | 1 - 2 | 147 | 101 |
BC137632.1 | - | 1 - 2 | 116 | 101 |